Bicyclo[3.2.0]hept-2-en-6-one ≥97%

Supplier: BeanTown Chemical

Synonyms: (±)-cis-Bicyclo[3.2.0]hept-2-en-6-one

135630-100G 135630-25G 135630-5G
BT135630-100GEA 1976 USD
BT135630-100G BT135630-25G BT135630-5G
Bicyclo[3.2.0]hept-2-en-6-one ≥97%
Bicyclo[3.2.0]hept-2-en-6-one

CAS: 13173-09-6; MDL No: MFCD00066201 UN No: UN1224; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C7H8O; MW: 108.14 Boiling Point: 158-160° Density (g/mL): 1.025; Refractive Index: 1.482 Air Sensitive


  • Refractive index: 1.482
  • Air Sensitive
  • Liquid

Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 158…160 °C
Density: 1.025 g/mL
MDL Number: MFCD00066201
CAS Number: 13173-09-6
UN: 1224
ADR: 3 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR