Ruthenium(III) acetylacetonate
Supplier: BeanTown Chemical
Synonyms:
Ruthenium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')ruthenium (III), Ruthenium (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato)ruthenium(III)
135580-1G
135580-5G
BT135580-1GEA
87
USD
BT135580-1G
BT135580-5G
Ruthenium(III) acetylacetonate
Ruthenium (III) acetylacetonate
CAS: 14284-93-6; EC No: 238-193-0; MDL No: MFCD00000030
Crystalline; Linear Formula: Ru(C5H7O2)3; MW: 398.40
Melting Point: 260° (decomposes)
- Crystalline
Formula:
C₁₅H₂₁O₆Ru Melting Pt: 260 °C (dec.) |
MDL Number:
MFCD00000030 CAS Number: 14284-93-6 EINECS: 238-193-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...