1-(2-Hydroxyethyl)pyrrolidine ≥97%
Supplier: BeanTown Chemical
Synonyms:
1-Pyrrolidineethanol, N-(2-Hydroxyethyl)pyrrolidine
135530-100ML
135530-25ML
BT135530-100MLEA
152
USD
BT135530-100ML
BT135530-25ML
1-(2-Hydroxyethyl)pyrrolidine ≥97%
1-(2-Hydroxyethyl)pyrrolidine
CAS: 2955-88-6; EC No: 220-976-3; MDL No: MFCD00003181; RTECS: UY1175000
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C6H13NO; MW: 115.17
Boiling Point: 79-81°/13 mmHg; Flash point: 56°C (133°F)
Density (g/mL): 0.985; Refractive Index: 1.471
- RTECS: UY1175000
- Refractive index: 1.471
- Liquid
Formula:
C₆H₁₃NO Boiling Pt: 79…81 °C/13 mmHg Density: 0.985 g/mL Flash Pt: 56 °C (133 °F) |
MDL Number:
MFCD00003181 CAS Number: 2955-88-6 EINECS: 220-976-3 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...