Lithium hydroxide monohydrate ≥98.0% ACS

Supplier: BeanTown Chemical

135445-100G
BT135445-100GEA 101.37 USD
BT135445-100G BT135445-2KG BT135445-500G
Lithium hydroxide monohydrate ≥98.0% ACS
Lithium hydroxide monohydrate

CAS: 1310-66-3; EC No: 215-183-4; MDL No: MFCD00149772; RTECS: OJ6307080 UN No: UN2680; Haz Class: 8; Packing Group: II Powder; Linear Formula: LiOH·H2O ; MW: 41.96 Density (g/mL): 1.51 Air Sensitive , Hygroscopic


  • RTECS: OJ6307080
  • Air Sensitive, Hygroscopic
  • Powder

Formula: LiOH·1H₂O
MW: 41.96 g/mol
Boiling Pt: 920
Melting Pt: 462
Density: 1.51 g/mL
MDL Number: MFCD00149772
CAS Number: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8 II
Merck Index: 13,05556

Order Now

Specification Test Results

Assay 98.0% min.
Insoluble matter ≤0.01%
Chloride (Cl) ≤0.01%
Sulfate (SO4) ≤0.05%
Heavy metals (as Pb) ≤0.002%
Iron (Fe) ≤0.002%
Lithium carbonate (Li2CO3) ≤2.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR