Copper(II) fluoride, anhydrous ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Cupric fluoride
135435-10G
135435-50G
BT135435-10GEA
73.18
USD
BT135435-10G
BT135435-50G
Copper(II) fluoride, anhydrous ≥99.5%
Copper (II) fluoride
CAS: 7789-19-7; EC No: 232-147-3; MDL No: MFCD00010977
UN No: UN3260; Haz Class: 8; Packing Group: III
Powder; Molecular Formula: CuF2; MW: 101.54
Melting Point: 950° (decomposes)
Density (g/mL): 4.23
Hygroscopic
- Hygroscopic
- Powder
Formula:
CuF₂ MW: 101.54 Boiling Pt: 950 Melting Pt: 855 (dec.) Density: 4.23 g/mL |
MDL Number:
MFCD00010977 CAS Number: 7789-19-7 EINECS: 232-147-3 UN: 3260 ADR: 6.1 III Merck Index: 12,02704 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...