Petroleum spirit, 35…60 °C ACS
Supplier: BeanTown Chemical
Synonyms:
Gasoline, Petroleum ether, Petrol, Benzine
135430-1L
135430-4L
135430-4X1L
BT135430-1LEA
54.39
USD
BT135430-1L
BT135430-4L
BT135430-4X1L
Petroleum spirit, 35…60 °C ACS
Petroleum spirit
CAS: 8032-32-4; EC No: 232-453-7; MDL No: MFCD00081849; RTECS: OI6180000
UN No: UN1268; Haz Class: 3; Packing Group: I
Liquid
Boiling Point: 35-60°; Flash point: -49°C (-57°F)
Density (g/mL): 0.64; Refractive Index: 1.363
- RTECS: OI6180000
- Refractive index: 1.363
- Liquid
Boiling Pt:
35…60 °C Density: 0.64 g/mL Flash Pt: –49 °C (–57 °F) |
MDL Number:
MFCD00081849 CAS Number: 64742-49-0 EINECS: 232-453-7 UN: 1268 ADR: 3,I |
Specification Test Results
Color (APHA) | ≤10 |
Boiling range, 35-60°C | Passes test |
Residue after evaporation | ≤10ppm |
Acidity | Passes test |
Copper Corrosion | Passes test |
Sulfur (S) | ≤5ppm |
Benzene (GC) | ≤2ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...