(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane ≥97%
Supplier: BeanTown Chemical
Synonyms:
tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
135410-1G
135410-250MG
135410-5G
BT135410-1GEA
180
USD
BT135410-1G
BT135410-250MG
BT135410-5G
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane ≥97%
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane
CAS: 113451-59-5; MDL No: MFCD01569250
Powder; Molecular Formula: C10H18N2O2; MW: 198.26
Melting Point: 74-76°
Optical Rotation: [α]22/D -44°, c = 1 in chloroform
Air Sensitive
- Optical rotation: [α]22/D -44°, c = 1 in chloroform
- Air Sensitive
- Powder
Formula:
C₁₀H₁₈N₂O₂ Melting Pt: 74…76 °C |
MDL Number:
MFCD01569250 CAS Number: 113451-59-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...