Cobalt(II) acetate tetrahydrate
Supplier: BeanTown Chemical
Synonyms:
Cobaltous acetate tetrahydrate, Acetic acid cobalt (II) salt tetrahydrate, Cobalt(Ⅱ) acetate tetrahydrate, Cobalt diacetate tetrahydrate
135395-1KG
135395-250G
BT135395-1KGEA
122.07
USD
BT135395-1KG
BT135395-250G
Cobalt(II) acetate tetrahydrate
Cobalt (II) acetate tetrahydrate
CAS: 6147-53-1; EC No: 200-755-8; MDL No: MFCD00149569; RTECS: AG3325000
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline; Linear Formula: (CH3CO2)2Co·4H2O; Molecular Formula: C4H6CoO4·4H2O; MW: 249.08
Hygroscopic
- RTECS: AG3325000
- Hygroscopic
- Crystalline
Formula:
(H₃CCOO)₂Co·4H₂O |
MDL Number:
MFCD00149569 CAS Number: 6147-53-1 EINECS: 200-755-8 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...