Valeric acid ≥99%

Supplier: BeanTown Chemical

Synonyms: Pentanoic acid

135390-100ML 135390-2.5L 135390-500ML
BT135390-100MLEA 27.73 USD
BT135390-100ML BT135390-2.5L BT135390-500ML
Valeric acid ≥99%
Valeric acid

CAS: 109-52-4; EC No: 203-677-2; MDL No: MFCD00004413; RTECS: YV6100000 UN No: UN3265; Haz Class: 8; Packing Group: III Liquid; Linear Formula: CH3(CH2)3COOH; Molecular Formula: C5H10O2; MW: 102.13 Melting Point: -20 - -18°; Boiling Point: 184-186°; Flash point: 88°C (190°F) Density (g/mL): 0.939; Refractive Index: 1.408


  • RTECS: YV6100000
  • Refractive index: 1.408
  • Liquid

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 186
Melting Pt: -32
Density: 0.939 g/mL
Flash Pt: 88 °C (190 °F)
MDL Number: MFCD00004413
CAS Number: 109-52-4
EINECS: 203-677-2
UN: 3265
ADR: 8 III
Merck Index: 12,10042

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR