Sodium hydroxide 2.0 N in water volumetric solution

Supplier: BeanTown Chemical

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

135350-1L 135350-4L
BT135350-1LEA 40.13 USD
BT135350-1L BT135350-4L
Sodium hydroxide 2.0 N in water volumetric solution
Sodium hydroxide

CAS: 1310-73-2; EC No: 215-185-5; MDL No: MFCD00003548 UN No: UN1824; Haz Class: 8; Packing Group: II Liquid; Linear Formula: NaOH; MW: 40.00 Melting Point: -12-10°; Boiling Point: 105-140° Density (g/mL): 1.08 Air Sensitive


  • Air Sensitive
  • Liquid

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.08 g/mL
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR