Potassium manganate
Supplier: BeanTown Chemical
135340-10G
135340-50G
BT135340-10GEA
69.33
USD
BT135340-10G
BT135340-50G
Potassium manganate
Potassium manganate
CAS: 10294-64-1; EC No: 233-665-2; MDL No: MFCD00043089
UN No: UN1479; Haz Class: 5.1; Packing Group: III
Powder; Molecular Formula: K2MnO4; MW: 197.13
Melting Point: 190°
- Powder
Formula:
K₂MnO₄ Melting Pt: 190 °C |
MDL Number:
MFCD00043089 CAS Number: 10294-64-1 EINECS: 233-665-2 UN: 1479 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...