Diethyl malonate ≥99%

Supplier: BeanTown Chemical

Synonyms: 1,3-Propanedioic acid diethyl ester, Malonic acid diethyl ester

135220-10KG 135220-2.5KG 135220-500G
BT135220-10KGEA 310 USD
BT135220-10KG BT135220-2.5KG BT135220-500G
Diethyl malonate ≥99%
Diethyl malonate

CAS: 105-53-3; EC No: 203-305-9; MDL No: MFCD00009195; RTECS: OO0700000 Liquid; Molecular Formula: C7H12O4; MW: 160.17 Melting Point: -50°; Boiling Point: 199°; Flash point: 90°C (194°F) Density (g/mL): 1.055; Refractive Index: 1.413


  • RTECS: OO0700000
  • Refractive index: 1.413
  • Liquid

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 199
Melting Pt: -50
Density: 1.055 g/mL
Flash Pt: 90 °C (194 °F)
MDL Number: MFCD00009195
CAS Number: 105-53-3
EINECS: 203-305-9
Merck Index: 13,03856

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR