1-Octanesulfonic acid sodium salt ≥99%
Supplier: BeanTown Chemical
Synonyms:
Sodium 1-octansulfonate, Sodium octane-1-sulphonate
135195-100G
135195-25G
135195-5G
BT135195-100GEA
544.69
USD
BT135195-100G
BT135195-25G
BT135195-5G
1-Octanesulfonic acid sodium salt ≥99%
1-Octanesulphonic acid sodium salt
CAS: 5324-84-5; EC No: 226-195-4; MDL No: MFCD00007544
Solid; Linear Formula: CH3(CH2)7SO3Na; MW: 216.28
Melting Point:
- Solid
Formula:
C₈H₁₇NaO₃S MW: 216.28 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00007544 CAS Number: 5324-84-5 EINECS: 226-195-4 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...