1-Octanesulfonic acid sodium salt ≥99%

Supplier: BeanTown Chemical

Synonyms: Sodium 1-octansulfonate, Sodium octane-1-sulphonate

135195-100G 135195-25G 135195-5G
BT135195-100GEA 544.69 USD
BT135195-100G BT135195-25G BT135195-5G
1-Octanesulfonic acid sodium salt ≥99%
1-Octanesulphonic acid sodium salt

CAS: 5324-84-5; EC No: 226-195-4; MDL No: MFCD00007544 Solid; Linear Formula: CH3(CH2)7SO3Na; MW: 216.28 Melting Point: 300°


  • Solid

Formula: C₈H₁₇NaO₃S
Melting Pt: >300 °C
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR