4-(Indol-3-yl)butyric acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Indolebutyric acid, Indole-3-butyric acid

135160-100G 135160-25G 135160-5G
BT135160-100GEA 369 USD
BT135160-100G BT135160-25G BT135160-5G
4-(Indol-3-yl)butyric acid ≥98%
4-(Indol-3-yl)butyric acid

CAS: 133-32-4; EC No: 205-101-5; MDL No: MFCD00005664; RTECS: NL5250000 UN No: UN2811 ; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Molecular Formula: C12H13NO2; MW: 203.24 Melting Point: 122-125° Air Sensitive


  • RTECS: NL5250000
  • Air Sensitive
  • Crystalline/Powder

Formula: C₁₂H₁₃NO₂
MW: 203.24
Melting Pt: 122…125 °C
Density: 1.27
MDL Number: MFCD00005664
CAS Number: 133-32-4
EINECS: 205-101-5
UN: 2811
ADR: 6.1 III
Merck Index: 13,04987

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR