4-(Indol-3-yl)butyric acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
3-Indolebutyric acid, Indole-3-butyric acid
135160-100G
135160-25G
135160-5G
BT135160-100GEA
369
USD
BT135160-100G
BT135160-25G
BT135160-5G
4-(Indol-3-yl)butyric acid ≥98%
4-(Indol-3-yl)butyric acid
CAS: 133-32-4; EC No: 205-101-5; MDL No: MFCD00005664; RTECS: NL5250000
UN No: UN2811 ; Haz Class: 6.1; Packing Group: III
Crystalline/Powder; Molecular Formula: C12H13NO2; MW: 203.24
Melting Point: 122-125°
Air Sensitive
- RTECS: NL5250000
- Air Sensitive
- Crystalline/Powder
Formula:
C₁₂H₁₃NO₂ Melting Pt: 122…125 °C |
MDL Number:
MFCD00005664 CAS Number: 133-32-4 EINECS: 205-101-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...