n-Nonadecane ≥99%
Supplier: BeanTown Chemical
Synonyms:
N-nonadecane
135135-100G
135135-25G
BT135135-100GEA
70.68
USD
BT135135-100G
BT135135-25G
n-Nonadecane ≥99%
n-Nonadecane
CAS: 629-92-5; EC No: 211-116-8; MDL No: MFCD00009012
Solid; Linear Formula: CH3(CH2)17CH3; Molecular Formula: C19H40; MW: 268.53
Melting Point: 30-34°; Boiling Point: 440°; Flash point: 100°C (212°F)
Density (g/mL): 0.786; Refractive Index: 1.441
- Refractive index: 1.441
- Solid
Formula:
H₃C(CH₂)₁₇CH₃ Boiling Pt: 440 °C Melting Pt: 30…34 °C Density: 0.786 g/mL Flash Pt: 100 °C (212 °F) |
MDL Number:
MFCD00009012 CAS Number: 629-92-5 EINECS: 211-116-8 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...