Silicon carbide, β-phase ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

135095-100G 135095-2KG 135095-500G
BT135095-100GEA 90 USD
BT135095-100G BT135095-2KG BT135095-500G
Silicon carbide, β-phase ≥99.5% (trace metals basis)
Silicon carbide, β-phase

CAS: 409-21-2; EC No: 206-991-8; MDL No: MFCD00049531 1 Micron Powder, S.A. 11.5 m2/g; Linear Formula: SiC; MW: 40.10 Melting Point: 2700° Density (g/mL): 3.22


  • 1 Micron Powder, S.A. 11.5 m²/g

Formula: SiC
Melting Pt: 2700 °C
Density: 3.22 g/mL
MDL Number: MFCD00049531
CAS Number: 409-21-2
EINECS: 206-991-8

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR