1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Catechol-3,5-disulphonic acid disodium salt monohydrate, 4,5-Dihydroxy-1,3-benzenedisulphonic acid disodium salt monohydrate, Disodium 4,5-dihydroxybenzene-1,3-disulfonate monohydrate, Tiron monohydrate, 3,5-Pyrocatecholdisulphonic acid disodium salt monohydrate, Disodium 4,5-dihydroxybenzene-1,3-disulphonate monohydrate, Catechol-3,5-disulfonic acid disodium salt monohydrate, 3,5-Pyrocatecholdisulfonic acid disodium salt monohydrate, 4,5-Dihydroxy-1,3-benzenedisulfonic acid disodium salt monohydrate
CAS: 270573-71-2; EC No: 205-741-5; MDL No: MFCD00007473; RTECS: CZ9263000
Solid; Molecular Formula: C6H4Na2O8S2·H2O; MW: 332.22
Melting Point:
- RTECS: CZ9263000
- Hygroscopic
- Solid
Formula:
C₆H₄Na₂O₈S₂·1H₂O Melting Pt: >300 °C |
MDL Number:
MFCD00149531 CAS Number: 270573-71-2 EINECS: 205-741-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...