Magnesium acetate 1 M in water
Supplier: BeanTown Chemical
135030-100ML
135030-50ML
BT135030-100MLEA
81.05
USD
BT135030-100ML
BT135030-50ML
Magnesium acetate 1 M in water
Magnesium acetate
CAS: 142-72-3; EC No: 205-554-9; MDL No: MFCD00149214; RTECS: AI5600000
Liquid; Linear Formula: (CH3COO)2Mg; MW: 142.39
Melting Point: 72-75°
Density (g/mL): 1.074; Refractive Index: 1.358
- RTECS: AI5600000
- Refractive index: 1.358
- Liquid
Formula:
(H₃CCOO)₂Mg Melting Pt: 72…75 °C Density: 1.074 g/mL |
MDL Number:
MFCD00003539 CAS Number: 142-72-3 EINECS: 205-554-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...