Silicon dioxide ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
134980-100G
134980-25G
BT134980-100GEA
196
USD
BT134980-100G
BT134980-25G
Silicon dioxide ≥99.9% (trace metals basis)
Silicium dioxide
CAS: 7631-86-9; EC No: 231-545-4; MDL No: MFCD00011232
1.5 Micron Powder; Linear Formula: SiO2; MW: 60.08
Melting Point: 1610°; Boiling Point: 2230°
Density (g/mL): 2.2-2.6; Refractive Index: 1.544
- Refractive index: 1.544
- 1.5 Micron Powder
Formula:
SiO₂ Boiling Pt: 2230 °C Melting Pt: 1610 °C Density: 2.2…2.6 g/mL |
MDL Number:
MFCD00011232 CAS Number: 7631-86-9 EINECS: 231-545-4 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...