Dichromium trisulfide ≥99%

Supplier: BeanTown Chemical

Synonyms: Chromium (III) sulphide, Chromium (III) sulfide

134885-25G 134885-5G
BT134885-25GEA 510 USD
BT134885-25G BT134885-5G
Dichromium trisulfide ≥99%
Dichromium trisulphide

CAS: 12018-22-3; EC No: 234-638-8; MDL No: MFCD00010950 -200 Mesh Powder; Molecular Formula: Cr2S3; MW: 200.18


  • -200 Mesh Powder

Formula: Cr₂S₃
MW: 200.19
Density: 3.77
MDL Number: MFCD00010950
CAS Number: 12018-22-3
EINECS: 234-638-8

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR