Imidazole ≥99%
Supplier: BeanTown Chemical
Synonyms:
Glyoxaline, 1H-Imidazole, Glyoxalamine
134805-100G
134805-2.5KG
134805-500G
BT134805-100GEA
36.73
USD
BT134805-100G
BT134805-2.5KG
BT134805-500G
Imidazole ≥99%
Imidazole
CAS: 288-32-4; EC No: 206-019-2; MDL No: MFCD00005183; RTECS: NI3325000
UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III
Crystalline; Molecular Formula: C3H4N2; MW: 68.08
Melting Point: 85-92°; Boiling Point: 256°; Flash point: 145°C (293°F)
- RTECS: NI3325000
- Crystalline
Formula:
C₃H₄N₂ Boiling Pt: 256 °C Melting Pt: 85…92 °C Flash Pt: 145 °C (293 °F) |
MDL Number:
MFCD00005183 CAS Number: 288-32-4 EINECS: 206-019-2 UN: 2923 ADR: 8,III |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...