Cyclopropylméthanol ≥98%

Supplier: BeanTown Chemical

Synonyms: Cyclopropyl carbinol, Cyclopropanemethanol

134780-100G 134780-25G 134780-5G
BT134780-100GEA 159 USD
BT134780-100G BT134780-25G BT134780-5G
Cyclopropylméthanol ≥98%
Cyclopropylméthanol

CAS: 2516-33-8; EC No: 219-735-5; MDL No: MFCD00001309; RTECS: GZ2212250 UN No: UN1987; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C4H8O; MW: 72.11 Boiling Point: 123-124°/738 mmHg; Flash point: 35°C (95°F) Density (g/mL): 0.89; Refractive Index: 1.431


  • RTECS: GZ2212250
  • Refractive index: 1.431
  • Liquid

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 123
Melting Pt: -60
Density: 0.89 g/mL
Flash Pt: 35 °C (95 °F)
MDL Number: MFCD00001309
CAS Number: 2516-33-8
EINECS: 219-735-5
UN: 1987
ADR: 3 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR