2,2-Bis(4-glycidyloxyphenyl)propane
Supplier: BeanTown Chemical
Synonyms:
Bisphenol A Diglycidyl Ether, 2,2-Bis(4-hydroxyphenyl)propane Diglycidyl Ether, DER 332, BADGE, D.E.R. 332 epoxy resin
134745-100G
134745-25G
134745-500G
BT134745-100GEA
72.02
USD
BT134745-100G
BT134745-25G
BT134745-500G
2,2-Bis(4-glycidyloxyphenyl)propane
2,2-Bis(4-glycidyloxyphenyl)propane
CAS: 1675-54-3; EC No: 216-823-5; MDL No: MFCD00080480; RTECS: TX3800000
Liquid; Molecular Formula: C21H24O4; MW: 340.41
Density (g/mL): 1.16
- RTECS: TX3800000
- Liquid
Formula:
C₂₁H₂₄O₄ MW: 340.42 g/mol Density: 1.16 g/mL |
MDL Number:
MFCD00080480 CAS Number: 1675-54-3 EINECS: 216-823-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...