4,6-Dihydroxypyrimidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-Hydroxy-1H-pyrimidin-4-one

134720-100G 134720-25G 134720-500G
BT134720-100GEA 138 USD
BT134720-100G BT134720-25G BT134720-500G
4,6-Dihydroxypyrimidine ≥98%
4,6-Dihydroxypyrimidine

CAS: 1193-24-4; EC No: 214-772-3; MDL No: MFCD00016733; RTECS: UW7523000 Powder; Molecular Formula: C4H4N2O2; MW: 112.09 Melting Point: 300°


  • RTECS: UW7523000
  • Powder

Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00016733
CAS Number: 1193-24-4
EINECS: 214-772-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR