1-Chlorobutane ≥99.5% for HPLC

Supplier: BeanTown Chemical

Synonyms: n-Butyl Chloride

134620-1L
BT134620-1LEA 84.09 USD
BT134620-1L BT134620-250ML BT134620-4L
1-Chlorobutane ≥99.5% for HPLC
1-Chlorobutane

CAS: 109-69-3; EC No: 203-696-6; MDL No: MFCD00001009; RTECS: EJ6300000 UN No: UN1127; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3(CH2)3Cl; Molecular Formula: C4H9Cl; MW: 92.57 Melting Point: -123°; Boiling Point: 77-78°; Flash point: -12°C (10°F) Density (g/mL): 0.886; Refractive Index: 1.402


  • RTECS: EJ6300000
  • Refractive index: 1.402
  • Liquid

Formula: CH₃(CH₂)₃Cl
MW: 92.57 g/mol
Boiling Pt: 77…78 °C
Melting Pt: –123 °C
Density: 0.886 g/mL
Flash Pt: –12 °C (10 °F)
MDL Number: MFCD00001009
CAS Number: 109-69-3
EINECS: 203-696-6
UN: 1127
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bd980eb6e7ecd91951d46d0ea510119e976dc0&vid=5231549172a609ad96623a80204b11c7c0c69273&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR