Isonicotinaldehyde ≥97%
Supplier: BeanTown Chemical
Synonyms:
Pyridine-4-carboxaldehyde, 4-Pyridinecarboxaldehyde, 4-Pyridinecarbaldehyde
134615-100G
134615-25G
134615-500G
BT134615-100GEA
103
USD
BT134615-100G
BT134615-25G
BT134615-500G
Isonicotinaldehyde ≥97%
Isonicotinaldehyde
CAS: 872-85-5; EC No: 212-832-3; MDL No: MFCD00006425; RTECS: NR9400000
Liquid; Molecular Formula: C6H5NO; MW: 107.11
Melting Point: -4°; Boiling Point: 198°; Flash point: 75°C (167°F)
Density (g/mL): 1.137; Refractive Index: 1.544
- RTECS: NR9400000
- Refractive index: 1.544
- Liquid
Formula:
C₆H₅NO Boiling Pt: 198 °C Melting Pt: –4 °C Density: 1.137 g/mL Flash Pt: 75 °C (167 °F) |
MDL Number:
MFCD00006425 CAS Number: 872-85-5 EINECS: 212-832-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...