Niobium(V) ethanolate ≥99.999%
Supplier: BeanTown Chemical
Synonyms:
Niobium(V) ethoxide
134610-1G
134610-5G
BT134610-1GEA
29
USD
BT134610-1G
BT134610-5G
Niobium(V) ethanolate ≥99.999%
Niobium(V) ethanolate
CAS: 3236-82-6; EC No: 221-795-2; MDL No: MFCD00015122
UN No: UN2924; Haz Class: 3(8); Packing Group: III
Liquid; Linear Formula: Nb(OCH2CH3)5; MW: 318.21
Melting Point: 6°; Boiling Point: 142°/0.1 mmHg; Flash point: 36°C (97°F)
Density (g/mL): 1.268; Refractive Index: 1.516
Air Sensitive, Moisture Sensitive
- Refractive index: 1.516
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₁₀H₂₅NbO₅ Boiling Pt: 142 °C/0.1 mmHg Melting Pt: 6 °C Density: 1.268 g/mL Flash Pt: 36 °C (97 °F) |
MDL Number:
MFCD00015122 CAS Number: 3236-82-6 EINECS: 221-795-2 ADR: 3(8),III |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...