Citric acid, anhydrous ≥99.5% ACS
Supplier: BeanTown Chemical
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
134485-100G
134485-2KG
134485-500G
BT134485-100GEA
25
USD
BT134485-100G
BT134485-2KG
BT134485-500G
Citric acid, anhydrous ≥99.5% ACS
Citric acid
CAS: 77-92-9; EC No: 201-069-1; MDL No: MFCD00011669; RTECS: GE7350000
Crystalline; Molecular Formula: C6H8O7; MW: 192.13
Melting Point: 153-159°
Hygroscopic
- RTECS: GE7350000
- Hygroscopic
- Crystalline
Formula:
HOC(COOH)(CH₂COOH)₂ MW: 192.13 g/mol Boiling Pt: 310 Melting Pt: 153…159 °C |
MDL Number:
MFCD00011669 CAS Number: 77-92-9 EINECS: 201-069-1 Merck Index: 13,02350 |
|
Specification Test Results
Assay | 99.5% |
Insoluble matter | ≤0.005% |
Chloride (Cl) | ≤0.001% |
Oxalate (C2O4) | ≤0.05% |
Phosphate (PO4) | ≤0.001% |
Sulfate (SO4) | ≤0.002% |
Iron (Fe) | ≤3ppm |
Lead (Pb) | ≤2ppm |
Residue after ignition | ≤0.02% |
Substances carbonizable by hot sulfuric acid | Passes test |
Tridodecylamine | ≤0.1mg/kg |
Water | ≤0.5% |
Clarity and color of solution | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...