Tris(4-methoxyphenyl)phosphine

Supplier: BeanTown Chemical

Synonyms: Tris(p-methoxyphenyl)phosphine

134475-1G 134475-25G 134475-5G
BT134475-1GEA 35 USD
BT134475-1G BT134475-25G BT134475-5G
Tris(4-methoxyphenyl)phosphine
Tris(4-methoxyphenyl)phosphine

CAS: 855-38-9; EC No: 212-723-0; MDL No: MFCD00014896 Powder; Linear Formula: (CH3OC6H4)3P; MW: 352.36 Melting Point: 131-134° Air Sensitive


  • Air Sensitive
  • Powder

Formula: C₂₁H₂₁O₃P
MW: 352.37 g/mol
Boiling Pt: 446
Melting Pt: 135
MDL Number: MFCD00014896
CAS Number: 855-38-9
EINECS: 212-723-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR