1,2-Epoxycyclohexane ≥98%

Supplier: BeanTown Chemical

Synonyms: Epoxycyclohexane, Cyclohexene oxide

134465-100G 134465-2.5KG 134465-500G
BT134465-100GEA 48 USD
BT134465-100G BT134465-2.5KG BT134465-500G
1,2-Epoxycyclohexane ≥98%
1,2-Epoxycyclohexane

CAS: 286-20-4; EC No: 206-007-7; MDL No: MFCD00005162; RTECS: RN7175000 UN No: UN2920; Haz Class: 8 (3); Packing Group: II Liquid; Molecular Formula: C6H10O; MW: 98.15 Boiling Point: 129-130°; Flash point: 24°C (75°F) Density (g/mL): 0.97; Refractive Index: 1.452


  • RTECS: RN7175000
  • Refractive index: 1.452
  • Liquid

Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 129
Melting Pt: -27
Density: 0.97 g/mL
Flash Pt: 24 °C (75 °F)
MDL Number: MFCD00005162
CAS Number: 286-20-4
EINECS: 206-007-7
UN: 2920
ADR: 8 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR