Hydrochlorothiazide ≥98%

Supplier: BeanTown Chemical

134455-100G 134455-25G 134455-5G
BT134455-100GEA 275.48 USD
BT134455-100G BT134455-25G BT134455-5G
Hydrochlorothiazide ≥98%
Hydrochlorothiazide

CAS: 58-93-5; EC No: 200-403-3; MDL No: MFCD00051765; RTECS: DK9100000 Solid; Molecular Formula: C7H8ClN3O4S2; MW: 297.74


  • RTECS: DK9100000
  • Solid

Formula: C₇H₈ClN₃O₄S₂
MDL Number: MFCD00051765
CAS Number: 58-93-5
EINECS: 200-403-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR