Diethyl methylphosphonite

Supplier: BeanTown Chemical

134385-10G 134385-2G 134385-500MG
BT134385-10GEA 531.2 USD
BT134385-10G BT134385-2G BT134385-500MG
Diethyl methylphosphonite
Diethyl methylphosphonite

CAS: 15715-41-0; EC No: 239-805-9; MDL No: MFCD00009089 UN No: UN3279; Haz Class: 6.1 (3); Packing Group: II Liquid; Linear Formula: (CH3CH2O)2PCH3; Molecular Formula: C5H13O2P; MW: 136.13 Flash point: 29°C (84°F) Density (g/mL): 0.907; Refractive Index: 1.419 Air Sensitive, Moisture Sensitive


  • Refractive index: 1.419
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₅H₁₃O₂P
MW: 136.13 g/mol
Density: 0.907 g/mL
Flash Pt: 29 °C (84 °F)
MDL Number: MFCD00009089
CAS Number: 15715-41-0
EINECS: 239-805-9
UN: 3279
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR