1,3-Diphenylguanidine ≥97%
Supplier: BeanTown Chemical
CAS: 102-06-7; EC No: 203-002-1; MDL No: MFCD00001758; RTECS: MF0875000
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline; Linear Formula: (C6H5NH)2C=NH; Molecular Formula: C13H13N3; MW: 211.26
Melting Point: 146-148°
Density (g/mL): 1.13
- RTECS: MF0875000
- Crystalline
Formula:
C₁₃H₁₃N₃ Melting Pt: 146…148 °C Density: 1.13 g/mL |
MDL Number:
MFCD00001758 CAS Number: 102-06-7 EINECS: 203-002-1 UN: 3077 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...