Gallium metal ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

134340-100G 134340-500G
BT134340-100GEA 348.75 USD
BT134340-100G BT134340-500G
Gallium metal ≥99.9% (trace metals basis)

CAS: 7440-55-3; EC No: 231-163-8; MDL No: MFCD00134045; RTECS: LW8600000 UN No: UN2803; Haz Class: 8; Packing Group: III Low Melting Solid; Molecular Formula: Ga; MW: 69.72 Melting Point: 29.8°; Boiling Point: 2403° Density (g/mL): 5.904


  • RTECS: LW8600000
  • Low Melting Solid

Formula: Ga
Boiling Pt: 2403 °C
Melting Pt: 29.8 °C
Density: 5.904 g/mL
MDL Number: MFCD00134045
CAS Number: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8,III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR