Diethyl diethylmalonate ≥98%

Supplier: BeanTown Chemical

134285-100G 134285-25G 134285-500G
BT134285-100GEA 130 USD
BT134285-100G BT134285-25G BT134285-500G
Diethyl diethylmalonate ≥98%
Diethyl diethylmalonate

CAS: 77-25-8; EC No: 201-016-2; MDL No: MFCD00009132 Liquid; Molecular Formula: C11H20O4 ; MW: 216.27 Boiling Point: 228-230°; Flash point: 94°C (201°F) Density (g/mL): 0.99; Refractive Index: 1.423


  • Refractive index: 1.423
  • Liquid

Formula: C₁₁H₂₀O₄
Boiling Pt: 228…230 °C
Density: 0.99 g/mL
Flash Pt: 94 °C (201 °F)
MDL Number: MFCD00009132
CAS Number: 77-25-8
EINECS: 201-016-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR