Zirconium carbide ≥99.5%
Supplier: BeanTown Chemical
134245-250G
134245-50G
BT134245-250GEA
279.76
USD
BT134245-250G
BT134245-50G
Zirconium carbide ≥99.5%
Zirconium carbide
CAS: 12070-14-3; EC No: 235-125-1; MDL No: MFCD00049640
UN No: UN3178; Haz Class: 4.1; Packing Group: III
-325 Mesh Powder; Linear Formula: ZrC; MW: 103.23
Melting Point: 3540°
Density (g/mL): 6.73
- -325 Mesh Powder
Formula:
CZr Melting Pt: 3540 °C Density: 6.73 g/mL |
MDL Number:
MFCD00049640 CAS Number: 12070-14-3 EINECS: 235-125-1 UN: 3178 ADR: 4.1,III |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...