Bismuth(III) nitrate basic

Supplier: BeanTown Chemical

Synonyms: Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate

134230-100G 134230-25G 134230-500G
BT134230-100GEA 58 USD
BT134230-100G BT134230-25G BT134230-500G
Bismuth(III) nitrate basic
Bismuth (III) nitrate basic

CAS: 1304-85-4; EC No: 215-136-8; MDL No: MFCD00064844; RTECS: EB2977000 UN No: UN1477; Haz Class: 5.1; Packing Group: II Powder; Linear Formula: Bi5O(OH)9(NO3)4 ; Molecular Formula: H9Bi5N4O22; MW: 1461.99 Hygroscopic


  • RTECS: EB2977000
  • Hygroscopic
  • Powder

Formula: Bi₅O(OH)₉(NO₃)₄
MDL Number: MFCD00064844
CAS Number: 1304-85-4
EINECS: 215-136-8
UN: 1477
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR