Trimethylphosphine ≥99%
Supplier: BeanTown Chemical
Synonyms:
Methyl phosphine
ampouled under argon
134115-1G
134115-25G
134115-5G
BT134115-1GEA
55.64
USD
BT134115-1G
BT134115-25G
BT134115-5G
Trimethylphosphine ≥99%
Trimethylphosphine
CAS: 594-09-2; EC No: 209-823-1; MDL No: MFCD00008510
UN No: UN2845; Haz Class: 4.2; Packing Group: I
Liquid; Linear Formula: (CH3)3P; MW: 76.08
Boiling Point: 38-40°; Flash point: -30°C (-22°F)
Density (g/mL): 0.738; Refractive Index: 1.428
Air Sensitive, Moisture Sensitive
- Refractive index: 1.428
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₃H₉P Boiling Pt: 38…40 °C Density: 0.738 g/mL Flash Pt: –30 °C (–22 °F) |
MDL Number:
MFCD00008510 CAS Number: 594-09-2 EINECS: 209-823-1 ADR: 4.2,I |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...