Starch 1%, Acculute™ standard volumetric solution, indicator Final Volume 1L

Supplier: BeanTown Chemical

134095-1UNIT 134095-6UNITS
BT134095-1UNITEA 42.47 USD
BT134095-1UNIT BT134095-6UNIT
Starch 1%, Acculute™ standard volumetric solution, indicator Final Volume 1L
Starch
Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00082026
CAS Number: 9005-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR