Uridine 5′-monophosphate disodium salt ≥99%
Supplier: BeanTown Chemical
Synonyms:
U 5′-P, Disodium uridine-5-monophosphate, Sodium ((2R,3S,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate, UMP, 5′-UMP-Na2, di-Sodium uridine-5-monophosphate, 5'-UMP 2Na, 5'-Uridylic acid disodium salt, Uridylic acid disodium salt, 5'-UMP disodium salt, Disodium UMP
134070-100G
134070-25G
134070-5G
BT134070-100GEA
821
USD
BT134070-100G
BT134070-25G
BT134070-5G
Uridine 5′-monophosphate disodium salt ≥99%
Uridine 5′-monophosphate disodium salt
CAS: 3387-36-8; EC No: 222-211-9; MDL No: MFCD00006525; RTECS: YU7975000
Solid; Molecular Formula: C9H11N2O9P; MW: 368.14
Melting Point: 209° (decomposes)
- RTECS: YU7975000
- Solid
Formula:
C₉H₁₁N₂Na₂O₉P MW: 368.15 g/mol Melting Pt: 209 °C (dec.) |
MDL Number:
MFCD00006525 CAS Number: 3387-36-8 EINECS: 222-211-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...