Triiodomethane ≥99%
Supplier: BeanTown Chemical
Synonyms:
Iodoform
134040-1KG
134040-250G
134040-50G
BT134040-1KGEA
431.14
USD
BT134040-1KG
BT134040-250G
BT134040-50G
Triiodomethane ≥99%
Triiodomethane
CAS: 75-47-8; EC No: 200-874-5; MDL No: MFCD00001069; RTECS: PB7000000
Solid; Molecular Formula: CHI3; MW: 393.73
Melting Point: 118-121°
Density (g/mL): 4.008
Light Sensitive
- RTECS: PB7000000
- Light Sensitive
- Solid
Formula:
CHI₃ Melting Pt: 118…121 °C Density: 4.008 g/mL |
MDL Number:
MFCD00001069 CAS Number: 75-47-8 EINECS: 200-874-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...