Triiodomethane ≥99%

Supplier: BeanTown Chemical

Synonyms: Iodoform

134040-1KG 134040-250G 134040-50G
BT134040-1KGEA 431.14 USD
BT134040-1KG BT134040-250G BT134040-50G
Triiodomethane ≥99%
Triiodomethane

CAS: 75-47-8; EC No: 200-874-5; MDL No: MFCD00001069; RTECS: PB7000000 Solid; Molecular Formula: CHI3; MW: 393.73 Melting Point: 118-121° Density (g/mL): 4.008 Light Sensitive


  • RTECS: PB7000000
  • Light Sensitive
  • Solid

Formula: CHI₃
Melting Pt: 118…121 °C
Density: 4.008 g/mL
MDL Number: MFCD00001069
CAS Number: 75-47-8
EINECS: 200-874-5

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR