Barium ethoxide 10% (w/v) in ethanol , 99.5% (trace metals basis)

Supplier: BeanTown Chemical

134010-25ML
BT134010-100MLEA 235.36 USD
BT134010-100ML BT134010-25ML
Barium ethoxide 10% (w/v) in ethanol , 99.5% (trace metals basis)
Barium ethoxide

CAS: 2914-19-4; EC No: 220-829-3; MDL No: MFCD00050488 UN No: UN3274; Haz Class: 3 (8); Packing Group: II Liquid; Linear Formula: Ba(OCH2CH3)2; MW: 227.42 Moisture Sensitive


  • Moisture Sensitive
  • Liquid

Formula: C₄H₁₀BaO₂
MW: 227.449 g/mol
MDL Number: MFCD00050488
CAS Number: 2914-19-4
EINECS: 220-829-3
ADR: 3 (8) II

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR