Niobium ≥99.5% (trace metals basis), rods Ø 6.35 mm, annealed

Supplier: BeanTown Chemical

133985-100CM 133985-25CM 133985-5CM
BT133985-100CMEA 455 USD
BT133985-100CM BT133985-25CM BT133985-5CM
Niobium ≥99.5% (trace metals basis), rods Ø 6.35 mm, annealed
Niobium

CAS: 7440-03-1; EC No: 231-113-5; MDL No: MFCD00011126 Powder; Molecular Formula: Nb ; MW: 92.91 Melting Point: 2468°; Boiling Point: 4742° Density (g/mL): 8.57


  • Powder

Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57 g/mL
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR