Diethyl phosphonate ≥96%
Supplier: BeanTown Chemical
133970-1KG
133970-250G
BT133970-1KGEA
216
USD
BT133970-1KG
BT133970-250G
Diethyl phosphonate ≥96%
Diethyl phosphonate
CAS: 762-04-9; EC No: 212-091-6; MDL No: MFCD00044573; RTECS: TG7875000
Liquid; Linear Formula: (CH3CH2O)2P(O)H; Molecular Formula: C4H11O3P; MW: 138.10
Boiling Point: 50-51°/2 mmHg; Flash point: 82°C (180°F)
Density (g/mL): 1.072; Refractive Index: 1.407
Moisture Sensitive
- RTECS: TG7875000
- Refractive index: 1.407
- Moisture Sensitive
- Liquid
Formula:
C₄H₁₁O₃P MW: 138.1 g/mol Boiling Pt: 50 Density: 1.072 g/mL Flash Pt: 82 °C (180 °F) |
MDL Number:
MFCD00044573 CAS Number: 762-04-9 EINECS: 212-091-6 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...