Tritolyl phosphate (mixture of isomers)

Supplier: BeanTown Chemical

133935-2.5L 133935-500ML
BT133935-2.5LEA 187.93 USD
BT133935-2.5L BT133935-500ML
Tritolyl phosphate (mixture of isomers)
Tritolyl phosphate (mixture of isomeres)

CAS: 1330-78-5; EC No: 215-548-8; MDL No: MFCD02102116; RTECS: TD0175000 UN No: UN2810; Haz Class: 6.1; Packing Group: II Liquid; Linear Formula: (CH3C6H4O)3PO; Molecular Formula: C21H21O4P; MW: 368.36 Melting Point: -40°; Boiling Point: 265°/10 mmHg; Flash point: 250°C (482°F) Density (g/mL): 1.143; Refractive Index: 1.555


  • RTECS: TD0175000
  • Refractive index: 1.555
  • Liquid

Formula: C₂₁H₂₁O₄P
Boiling Pt: 265 °C/10 mmHg
Melting Pt: <–40 °C
Density: 1.143 g/mL
Flash Pt: 250 °C (482 °F)
MDL Number: MFCD02102116
CAS Number: 1330-78-5
EINECS: 215-548-8
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR