(R)-(+)-2-Pyrrolidinone-5-carboxylic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: D-Pyr-OH, (R)-(+)-2-Pyrrolidone-5-carboxylic acid

133885-100G 133885-25G 133885-5G
BT133885-100GEA 848 USD
BT133885-100G BT133885-25G BT133885-5G
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid ≥98%
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid

CAS: 4042-36-8; EC No: 223-735-0; MDL No: MFCD00066212 Crystalline; Molecular Formula: C5H7NO3 ; MW: 129.11 Melting Point: 155-156° Optical Rotation: [α]22/D +10°, c = 1.5 in water


  • Optical rotation: [α]22/D +10°, c = 1.5 in water
  • Crystalline

Formula: C₅H₇NO₃
Melting Pt: 155…156 °C
MDL Number: MFCD00066212
CAS Number: 4042-36-8
EINECS: 223-735-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR