Ammonium sulfate ≥99.999%

Supplier: BeanTown Chemical

Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

133850-10G 133850-250G 133850-50G
BT133850-10GEA 29 USD
BT133850-10G BT133850-250G BT133850-50G
Ammonium sulfate ≥99.999%
Ammonium sulphate

CAS: 7783-20-2; EC No: 231-984-1; MDL No: MFCD00003391; RTECS: BS4500000 Crystalline; Linear Formula: (NH4)2SO4; MW: 132.14 Melting Point: 280° (decomposes) Density (g/mL): 1.77


  • RTECS: BS4500000
  • Crystalline

Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: 280 °C (dec.)
Density: 1.77 g/mL
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
Merck Index: 13,00559

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR